(S)-α-(2-Oxiranyl)anisol 95%

Lieferant: Apollo Scientific

Synonyme: (S)-Glycidyl phenyl ether

OR917934-5G
APOSOR917934-5GEA 95.7 EUR
APOSOR917934-5G
(S)-α-(2-Oxiranyl)anisol 95%
(S)-α-(2-Oxiranyl)anisol
Formel: C₉H₁₀O₂
Molecular Weight: 150.1745 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06659026
CAS-Nummer: 71031-03-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR