4-Oxo-1-phenyl-cyclohexanecarboxylic acid 97%

Lieferant: Apollo Scientific

OR917952-1G OR917952-250MG
APOSOR917952-1GEA 654 EUR
APOSOR917952-1G APOSOR917952-250MG
4-Oxo-1-phenyl-cyclohexanecarboxylic acid 97%
4-Oxo-1-phenyl-cyclohexanecarboxylic acid
Formel: C₁₃H₁₄O₃
Molecular Weight: 218.248 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06208311
CAS-Nummer: 75945-91-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR