Fmoc-L-Arg(Me)2(Pbf)-OH

Lieferant: Apollo Scientific
Achtung

Synonyme: (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-(((dimethylamino)((2,2,4,6,7-pentamethyl-2,3-dihydrobenzofuran)-5-sulphonamido)methylene)amino)pentanoic acid

OR917976-1G OR917976-250MG
APOSOR917976-1GEA 864 EUR
APOSOR917976-1G APOSOR917976-250MG
Fmoc-L-Arg(Me)2(Pbf)-OH
Fmoc-L-Arg(Me)2(Pbf)-OH
Formel: C₃₆H₄₄N₄O₇S
Molecular Weight: 676.822 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD15141977
CAS-Nummer: 1185841-84-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR