2-Amino-6-chloro-5-nitrotoluene 95%

Lieferant: Apollo Scientific

OR918001-5G
APOSOR918001-5GEA 174 EUR
APOSOR918001-5G
2-Amino-6-chloro-5-nitrotoluene 95%
2-Amino-6-chloro-5-nitrotoluene
Formel: C₇H₇ClN₂O₂
Molecular Weight: 186,596 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09026958
CAS-Nummer: 64863-10-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR