1-Imidazo[1,2-a]pyridin-2-ylmethanamine dihydrochloride 95%

Lieferant: Apollo Scientific
Achtung

OR918033-1G OR918033-250MG
APOSOR918033-1GEA 0 EUR
APOSOR918033-1G APOSOR918033-250MG
1-Imidazo[1,2-a]pyridin-2-ylmethanamine dihydrochloride 95%
1-Imidazo[1,2-a]pyridin-2-ylmethanamine dihydrochloride
Formel: C₈H₉N₃·2ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD04448842
CAS-Nummer: 452967-56-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR