Methyl-4-bromo-1-methyl-1H-pyrazole-3-carboxylate 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 4-bromo-1-methyl-1H-pyrazole-3-carboxylate

OR918037-5G
APOSOR918037-5GEA 125 EUR
APOSOR918037-5G
Methyl-4-bromo-1-methyl-1H-pyrazole-3-carboxylate 98%
Methyl-4-bromo-1-methyl-1H-pyrazole-3-carboxylate
Formel: C₆H₇BrN₂O₂
Molecular Weight: 219.04 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02253785
CAS-Nummer: 211738-66-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR