7-Oxa-4-azaspiro[2.5]octane hydrochloride 98%
Lieferant: Apollo Scientific
OR918050-100MG
APOSOR918050-100MGEA
548
EUR
APOSOR918050-100MG
7-Oxa-4-azaspiro[2.5]octane hydrochloride 98%
7-Oxa-4-azaspiro[2.5]octane hydrochloride
Formel:
C₆H₁₁NO·ClH Molecular Weight: 149,619 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD18914346 CAS-Nummer: 218595-22-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...