1-(2-Hydroxyethyl)piperidine-4-carbonitrile 95%

Lieferant: Apollo Scientific

OR918054-1G
APOSOR918054-1GEA 122 EUR
APOSOR918054-1G
1-(2-Hydroxyethyl)piperidine-4-carbonitrile 95%
1-(2-Hydroxyethyl)piperidine-4-carbonitrile
Formel: C₈H₁₄N₂O
Molecular Weight: 154,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18373979
CAS-Nummer: 21168-73-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR