2-Oxo-2H-chromene-6-carboxylic acid 95%

Lieferant: Apollo Scientific

OR918069-5G
APOSOR918069-5GEA 299 EUR
APOSOR918069-5G
2-Oxo-2H-chromene-6-carboxylic acid 95%
2-Oxo-2H-chromene-6-carboxylic acid
Formel: C₁₀H₆O₄
Molecular Weight: 190.16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06203567
CAS-Nummer: 7734-80-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR