Geranyl nitrile [mixture of (E) and (Z) isomers, (1:1)] 95%

Lieferant: Apollo Scientific

OR918075-25G
APOSOR918075-25GEA 132 EUR
APOSOR918075-25G
Geranyl nitrile [mixture of (E) and (Z) isomers, (1:1)] 95%
Geranyl nitrile [mixture of (E) and (Z) isomers, (1:1)]
Formel: C₁₀H₁₅N
Lagertemperatur: Raumtemperatur
MDL: MFCD00001926
CAS-Nummer: 5146-66-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR