1-(4-Amino-2-methoxy-phenyl)-ethanone 95%

Lieferant: Apollo Scientific

Synonyme: 1-(4-Amino-2-methoxyphenyl)ethanone

OR918133-1G
APOSOR918133-1GEA 40.6 EUR
APOSOR918133-1G
1-(4-Amino-2-methoxy-phenyl)-ethanone 95%
1-(4-Amino-2-methoxy-phenyl)-ethanone
Formel: C₉H₁₁NO₂
Molecular Weight: 165.189 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11848247
CAS-Nummer: 60207-18-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR