2,3-Dimethylpyridin-4-amine 95%

Lieferant: Apollo Scientific

Synonyme: 4-Amino-2,3-dimethylpyridine

OR918177-1G
APOSOR918177-1GEA 120 EUR
APOSOR918177-1G
2,3-Dimethylpyridin-4-amine 95%
2,3-Dimethylpyridin-4-amine
Formel: C₇H₁₀N₂
Molecular Weight: 122,168 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00030819
CAS-Nummer: 122475-57-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR