4-tert-Butyl-1H-pyrazole 98%

Lieferant: Apollo Scientific

Synonyme: 4-(tert-Butyl)-1H-pyrazole

OR918189-1G
APOSOR918189-1GEA 151 EUR
APOSOR918189-1G
4-tert-Butyl-1H-pyrazole 98%
4-tert-Butyl-1H-pyrazole
Formel: C₇H₁₂N₂
Molecular Weight: 124.184 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11847492
CAS-Nummer: 105285-21-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR