1-(3-Bromphenyl)pyrrolidin 97%

Lieferant: Apollo Scientific
Achtung

OR9182-250MG
APOSOR9182-250MGEA 0 EUR
APOSOR9182-250MG
1-(3-Bromphenyl)pyrrolidin 97%
1-(3-Bromphenyl)pyrrolidin
Formel: C₁₀H₁₂BrN
Siedepunkt: 76 °C/0,1mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD04112531
CAS-Nummer: 219928-13-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR