N-(6-Chloro-2-pyridinyl)-N,N-dimethylamine 98%
Lieferant: Apollo Scientific
Synonyme:
6-Chloro-N,N-dimethylpyridin-2-amine
OR918240-1G
APOSOR918240-1GEA
734
EUR
APOSOR918240-1G
N-(6-Chloro-2-pyridinyl)-N,N-dimethylamine 98%
N-(6-Chloro-2-pyridinyl)-N,N-dimethylamine
Formel:
C₇H₉ClN₂ Molecular Weight: 156,62 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13188973 CAS-Nummer: 1060801-42-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...