N-Ethyl-N-(2-pyrrolidinylmethyl)ethanamine 95%

Lieferant: Apollo Scientific

OR918256-1G
APOSOR918256-1GEA 0 EUR
APOSOR918256-1G
N-Ethyl-N-(2-pyrrolidinylmethyl)ethanamine 95%
N-Ethyl-N-(2-pyrrolidinylmethyl)ethanamine
Formel: C₉H₂₀N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06653367
CAS-Nummer: 121053-95-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR