Chloride ionophore I 97%

Lieferant: Apollo Scientific

OR918293-100MG OR918293-1G OR918293-250MG
APOSOR918293-100MGEA 0 EUR
APOSOR918293-100MG APOSOR918293-1G APOSOR918293-250MG
Chloride ionophore I 97%
Chloride ionophore I
Formel: C₄₄H₂₈ClMnN₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00012153
CAS-Nummer: 32195-55-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR