4-(Pyridin-2-yl)butan-1-ol 95%

Lieferant: Apollo Scientific

OR918342-1G
APOSOR918342-1GEA 168 EUR
APOSOR918342-1G
4-(Pyridin-2-yl)butan-1-ol 95%
4-(Pyridin-2-yl)butan-1-ol
Formel: C₉H₁₃NO
Molecular Weight: 151,206 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12828284
CAS-Nummer: 17945-79-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR