2-Pentyl-2-cyclopenten-1-on 95%
Lieferant: Apollo Scientific
OR918346-25G
OR918346-5G
APOSOR918346-25GEA
313
EUR
APOSOR918346-25G
APOSOR918346-5G
2-Pentyl-2-cyclopenten-1-on 95%
2-Pentyl-2-cyclopenten-1-one
Formel:
C₁₀H₁₆O Molecular Weight: 152.24 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00036631 CAS-Nummer: 25564-22-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...