2-Pentyl-2-cyclopenten-1-on 95%

Lieferant: Apollo Scientific

OR918346-25G OR918346-5G
APOSOR918346-25GEA 313 EUR
APOSOR918346-25G APOSOR918346-5G
2-Pentyl-2-cyclopenten-1-on 95%
2-Pentyl-2-cyclopenten-1-one
Formel: C₁₀H₁₆O
Molecular Weight: 152.24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00036631
CAS-Nummer: 25564-22-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR