N,N-Dimethyl-His-OMe 95%

Lieferant: Apollo Scientific

OR918382-1G OR918382-250MG
APOSOR918382-1GEA 864 EUR
APOSOR918382-1G APOSOR918382-250MG
N,N-Dimethyl-His-OMe 95%
N,N-Dimethyl-His-OMe
Formel: C₉H₁₅N₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD01318773
CAS-Nummer: 170227-64-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR