Methyl (4s)-trans-2,2,5-trimethyl-1,3-dioxolane-4-carboxylate 95%

Lieferant: Apollo Scientific

Synonyme: (4S,5R)-Methyl 2,2,5-trimethyl-1,3-dioxolane-4-carboxylate

OR918390-1G
APOSOR918390-1GEA 120 EUR
APOSOR918390-1G
Methyl (4s)-trans-2,2,5-trimethyl-1,3-dioxolane-4-carboxylate 95%
Methyl (4s)-trans-2,2,5-trimethyl-1,3-dioxolane-4-carboxylate
Formel: C₈H₁₄O₄
Molecular Weight: 174,194 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00077933
CAS-Nummer: 38410-80-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR