3-(Chloromethyl)-1,5-dimethyl-1H-pyrazole 97%
Lieferant: Apollo Scientific
|
Gefahr
|
OR9184-1G
APOSOR9184-1GEA
284
EUR
APOSOR9184-1G
3-(Chloromethyl)-1,5-dimethyl-1H-pyrazole 97%
3-(Chloromethyl)-1,5-dimethyl-1H-pyrazole
Formel:
C₆H₉ClN₂ Molecular Weight: 144,602 g/mol Schmelzpunkt: 63…66 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08060499 CAS-Nummer: 180519-09-9 UN: 1759 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...