4-Methyl-2-(2-methyl-1-propen-1-yl)tetrahydro-2H-pyran (cis/trans-Gemisch) 95%

Lieferant: Apollo Scientific

OR918409-25G OR918409-5G
APOSOR918409-25GEA 197 EUR
APOSOR918409-25G APOSOR918409-5G
4-Methyl-2-(2-methyl-1-propen-1-yl)tetrahydro-2H-pyran (cis/trans-Gemisch) 95%
4-Methyl-2-(2-methyl-1-propenyl)tetrahydropyran (cis- and trans- mixture)
Formel: C₁₀H₁₈O
Lagertemperatur: Raumtemperatur
MDL: MFCD00036607
CAS-Nummer: 16409-43-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR