2-(2,3-Dichlorphenyl)ethanol 96%

Lieferant: Apollo Scientific
Achtung

OR918460-5G
APOSOR918460-5GEA 56.7 EUR
APOSOR918460-5G
2-(2,3-Dichlorphenyl)ethanol 96%
2-(2,3-Dichlorphenyl)ethanol
Formel: C₈H₈Cl₂O
Molecular Weight: 191.06 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04112742
CAS-Nummer: 114686-85-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR