Methyl 2-amino-4-methoxybenzoate 98%
Lieferant: Apollo Scientific
Synonyme:
Methyl 2-amino-4-methoxylbenzoate
OR918474-1G
APOSOR918474-1GEA
0
EUR
APOSOR918474-1G
Methyl 2-amino-4-methoxybenzoate 98%
Methyl 2-amino-4-methoxybenzoate
Formel:
C₉H₁₁NO₃ Molecular Weight: 181.189 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06658470 CAS-Nummer: 50413-30-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...