6-Methoxy-2(1H)-pyridinone 95%

Lieferant: Apollo Scientific

OR918492-1G
APOSOR918492-1GEA 451 EUR
APOSOR918492-1G
6-Methoxy-2(1H)-pyridinone 95%
6-Methoxy-2(1H)-pyridinone
Formel: C₆H₇NO₂
Molecular Weight: 125,125 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15143445
CAS-Nummer: 22385-36-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR