N-Methyl-3-Chloropropylamin-Hydrochlorid 95%

Lieferant: Apollo Scientific

OR918572-1G OR918572-5G
APOSOR918572-1GEA 0 EUR
APOSOR918572-1G APOSOR918572-5G
N-Methyl-3-Chloropropylamin-Hydrochlorid 95%
N-Methyl-3-Chloropropylamin-Hydrochlorid
Formel: C₄H₁₀ClN
Lagertemperatur: Raumtemperatur
MDL: MFCD00144935
CAS-Nummer: 97145-88-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR