tert-Butyl-4-[4-(aminomethyl)phenyl]tetrahydro-1(2H)-pyrazinecarboxylate 90%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 4-(4-BOC-Pyrazino)benzylamine, 4-[(Aminomethyl)phenyl]piperazine, N1-BOC protected, tert-Butyl 4-(4-(aminomethyl)phenyl)piperazine-1-carboxylate

OR9186-1G
APOSOR9186-1GEA 193 EUR
APOSOR9186-1G
tert-Butyl-4-[4-(aminomethyl)phenyl]tetrahydro-1(2H)-pyrazinecarboxylate 90%
tert-Butyl-4-[4-(aminomethyl)phenyl]tetrahydro-1(2H)-pyrazinecarboxylate
Formel: C₁₆H₂₅N₃O₂
Molecular Weight: 291,4 g/mol
Schmelzpunkt: 83…87 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03791213
CAS-Nummer: 852180-47-3
UN: 2923
ADR: 8+6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR