2-tert-Butyl-1,3,4-oxadiazole 95%
Lieferant: Apollo Scientific
OR918655-1G
APOSOR918655-1GEA
647
EUR
APOSOR918655-1G
2-tert-Butyl-1,3,4-oxadiazole 95%
2-tert-Butyl-1,3,4-oxadiazole
Formel:
C₆H₁₀N₂O Molecular Weight: 126,156 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08689904 CAS-Nummer: 251540-53-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...