7-Methoxy-4-methylquinoline 95%

Lieferant: Apollo Scientific

OR918676-500MG
APOSOR918676-500MGEA 219 EUR
APOSOR918676-500MG
7-Methoxy-4-methylquinoline 95%
7-Methoxy-4-methylquinoline
Formel: C₁₁H₁₁NO
Lagertemperatur: Raumtemperatur
MDL: MFCD11655579
CAS-Nummer: 6238-12-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR