2,5-Dimethyl-4,6-dihydroxypyrimidine 95%

Lieferant: Apollo Scientific

Synonyme: 4(1H)-Pyrimidinone,6-hydroxy-2,5-dimethyl-

OR918718-1G
APOSOR918718-1GEA 50.9 EUR
APOSOR918718-1G
2,5-Dimethyl-4,6-dihydroxypyrimidine 95%
2,5-Dimethyl-4,6-dihydroxypyrimidine
Formel: C₆H₈N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD02685061
CAS-Nummer: 1194-74-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR