2-(2-Methoxyphenyl)ethanol 97%

Lieferant: Apollo Scientific

OR918735-10G OR918735-5G
APOSOR918735-10GEA 88.6 EUR
APOSOR918735-10G APOSOR918735-5G
2-(2-Methoxyphenyl)ethanol 97%
2-(2-Methoxyphenyl)ethanol
Formel: C₉H₁₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00004512
CAS-Nummer: 7417-18-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR