8-Oxa-3-azabicyclo[3.2.1]octane hydrochloride 97%
Lieferant: Apollo Scientific
OR918784-1G
OR918784-5G
APOSOR918784-1GEA
104
EUR
APOSOR918784-1G
APOSOR918784-5G
8-Oxa-3-azabicyclo[3.2.1]octane hydrochloride 97%
8-Oxa-3-azabicyclo[3.2.1]octane hydrochloride
Formel:
C₆H₁₂ClNO Molecular Weight: 149.62 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09800611 CAS-Nummer: 54745-74-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...