1-(2-Aminophenyl)piperazin 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR9188-250MG
APOSOR9188-250MGEA
0
EUR
APOSOR9188-250MG
1-(2-Aminophenyl)piperazin 95%
1-(2-Aminophenyl)piperazin
Formel:
C₁₀H₁₅N₃ Schmelzpunkt: 114…115 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03444413 CAS-Nummer: 13339-02-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...