(6-Chloro-1H-benzimidazol-2-yl)methanol 95%

Lieferant: Apollo Scientific

Synonyme: (6-Chloro-1H-benzo[d]imidazol-2-yl)methanol

OR918810-1G
APOSOR918810-1GEA 26.1 EUR
APOSOR918810-1G
(6-Chloro-1H-benzimidazol-2-yl)methanol 95%
(6-Chloro-1H-benzimidazol-2-yl)methanol
Formel: C₈H₇ClN₂O
Molecular Weight: 182,61 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09971879
CAS-Nummer: 6953-65-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR