Methyl 3-(1-aminoethyl)benzoate hydrochloride 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR918899-250MG
APOSOR918899-250MGEA
88.6
EUR
APOSOR918899-250MG
Methyl 3-(1-aminoethyl)benzoate hydrochloride 95%
Methyl 3-(1-aminoethyl)benzoate hydrochloride
Formel:
C₁₀H₁₃NO₂·ClH Lagertemperatur: Raumtemperatur |
MDL:
MFCD18432494 CAS-Nummer: 1263378-68-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...