(Methylthio)acetaldehyddimethylacetal 95%

Lieferant: Apollo Scientific

Synonyme: (Methylthio)acetaldehydedimethylacetal

OR918956-25G
APOSOR918956-25GEA 88.6 EUR
APOSOR918956-25G
(Methylthio)acetaldehyddimethylacetal 95%
(Methylthio)acetaldehyddimethylacetal
Formel: C₅H₁₂O₂S
Molecular Weight: 136.21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00009848
CAS-Nummer: 40015-15-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR