2,6-Dimethyl-4-methoxybenzoic acid 95%

Lieferant: Apollo Scientific

OR919035-1G
APOSOR919035-1GEA 81.2 EUR
APOSOR919035-1G
2,6-Dimethyl-4-methoxybenzoic acid 95%
2,6-Dimethyl-4-methoxybenzoic acid
Formel: C₁₀H₁₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD01194287
CAS-Nummer: 37934-89-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR