4-Brom-2-methylpyridin 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR9191-100G
OR9191-25G
OR9191-5G
OR9191-1G
APOSOR9191-100GEA
190
EUR
APOSOR9191-100G
APOSOR9191-25G
APOSOR9191-5G
APOSOR9191-1G
4-Brom-2-methylpyridin 98%
4-Brom-2-methylpyridin
Formel:
C₆H₆BrN Molecular Weight: 172.02 g/mol Dichte: 1.45 g/cm³ (25 °C) Flammpunkt: 78.9 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03788196 CAS-Nummer: 22282-99-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...