2,6-Dimethyl-p-benzochinon 95%
Lieferant: Apollo Scientific
Synonyme:
2,6-Dimethylcyclohexa-2,5-diene-1,4-dione
OR919100-5G
APOSOR919100-5GEA
63.8
EUR
APOSOR919100-5G
2,6-Dimethyl-p-benzochinon 95%
2,6-Dimethyl-p-benzochinon
Formel:
C₈H₈O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001605 CAS-Nummer: 527-61-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...