Quinoline-6-carboxamide 95%
Lieferant: Apollo Scientific
OR919127-1G
APOSOR919127-1GEA
209
EUR
APOSOR919127-1G
Quinoline-6-carboxamide 95%
Quinoline-6-carboxamide
Formel:
C₁₀H₈N₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD01312760 CAS-Nummer: 5382-43-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...