5-Bromo-1H-pyrrolo[2,3-c]pyridin-2(3H)-one 95+%
Lieferant: Apollo Scientific
|
Achtung
|
OR919134-250MG
APOSOR919134-250MGEA
137
EUR
APOSOR919134-250MG
5-Bromo-1H-pyrrolo[2,3-c]pyridin-2(3H)-one 95+%
5-Bromo-1H-pyrrolo[2,3-c]pyridin-2(3H)-one
Formel:
C₇H₅BrN₂O Molecular Weight: 213,031 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD11848386 CAS-Nummer: 1217002-90-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...