4-Chloro-2-(methylthio)pyrimidine-5-carbaldehyde 95%

Lieferant: Apollo Scientific

Synonyme: 4-Chloro-2-(methylsulphanyl)pyrimidine-5-carbaldehyde

OR919181-1G
APOSOR919181-1GEA 30.6 EUR
APOSOR919181-1G
4-Chloro-2-(methylthio)pyrimidine-5-carbaldehyde 95%
4-Chloro-2-(methylthio)pyrimidine-5-carbaldehyde
Formel: C₆H₅ClN₂OS
Molecular Weight: 188.635 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16657918
CAS-Nummer: 148256-82-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR