2-(1-Naphthyloxy)acetohydrazide 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(Naphthoxy)acetic acid hydrazide, 2-(Naphthalen-1-yloxy)acetohydrazide

OR9192-1G OR9192-5G
APOSOR9192-1GEA 21.9 EUR
APOSOR9192-1G APOSOR9192-5G
2-(1-Naphthyloxy)acetohydrazide 98%
2-(1-Naphthyloxy)acetohydrazide
Formel: C₁₂H₁₂N₂O₂
Molecular Weight: 216.24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00156157
CAS-Nummer: 24310-15-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR