2,6-Dichlorpyrimidin-4-carbonitril 97%

Lieferant: Apollo Scientific

OR919202-1G
APOSOR919202-1GEA 45.1 EUR
APOSOR919202-1G
2,6-Dichlorpyrimidin-4-carbonitril 97%
2,6-Dichlorpyrimidin-4-carbonitril
Formel: C₅HCl₂N₃
Molecular Weight: 173.9900 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16988336
CAS-Nummer: 26293-93-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR