Ethyl 2-ethylpyrimidine-5-carboxylate 95%

Lieferant: Apollo Scientific

OR919228-1G
APOSOR919228-1GEA 400 EUR
APOSOR919228-1G
Ethyl 2-ethylpyrimidine-5-carboxylate 95%
Ethyl 2-ethylpyrimidine-5-carboxylate
Formel: C₉H₁₂N₂O₂
Molecular Weight: 180,204 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17190092
CAS-Nummer: 72790-13-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR