N,N-Dimethylbenzamid 95%
Lieferant: Apollo Scientific
OR919250-100G
APOSOR919250-100GEA
85.7
EUR
APOSOR919250-100G
N,N-Dimethylbenzamid 95%
N,N-Dimethylbenzamid
Formel:
C₉H₁₁NO Molecular Weight: 149,19 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008320 CAS-Nummer: 611-74-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...