2-(2-Furyl)-1,3-dioxolan 98%

Lieferant: Apollo Scientific

Synonyme: Furfural Ethylene Acetal, 2-(2-Furyl)-1,3-dioxolane

OR919261-25G
APOSOR919261-25GEA 224 EUR
APOSOR919261-25G
2-(2-Furyl)-1,3-dioxolan 98%
2-(1,3-Dioxolan-2-yl)furan
Formel: C₇H₈O₃
Molecular Weight: 140.14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00671520
CAS-Nummer: 1708-41-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR