1-Methyl-1-(m-tolyl)hydrazine 95%
Lieferant: Apollo Scientific
OR919277-5G
APOSOR919277-5GEA
435
EUR
APOSOR919277-5G
1-Methyl-1-(m-tolyl)hydrazine 95%
1-Methyl-1-(m-tolyl)hydrazine
Formel:
C₈H₁₂N₂ Molecular Weight: 136,20 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03844802 CAS-Nummer: 72233-91-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...